Assay Details
Gene Symbol: adbp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064038.8 | NP_496439.1 | 3 - 4 | 295 | 68 | |
GenBank mRNA | AF303256.1 | - | 3 - 4 | 295 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
