Assay Details
Gene Symbol: sfxn-1.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267379.3 | NP_001254308.1 | 2 - 3 | 572 | 75 | |
NM_001373796.2 | NP_001359739.1 | 1 - 2 | 81 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001373796.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
