Assay Details
Gene Symbol: rgs-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267421.4 | NP_001254350.3 | 2 - 3 | 237 | 72 | |
NM_001373807.2 | NP_001360477.1 | 2 - 3 | 272 | 72 | ||
NM_001377886.2 | NP_001364555.1 | 4 - 5 | 446 | 72 | ||
NM_001381631.1 | NP_001368098.1 | 1 - 2 | 45 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
