Assay Details
Gene Symbol: acl-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064324.9 | NP_496725.2 | 8 - 9 | 1808 | 83 | |
GenBank mRNA | AJ414404.1 | - | 8 - 9 | 1803 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
