Assay Details
Gene Symbol: mes-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064591.6 | NP_496992.3 | 4 - 5 | 1612 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
