Assay Details
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267509.3 | NP_001254438.1 | 10 - 11 | 2253 | 104 | |
NM_001377905.1 | NP_001364664.1 | 43 - 44 | 12010 | 104 | ||
NM_064643.10 | NP_497044.3 | 34 - 35 | 9750 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
