Assay Details
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267509.3 | NP_001254438.1 | 5 - 6 | 938 | 74 | |
NM_001377905.1 | NP_001364664.1 | 38 - 39 | 10695 | 74 | ||
NM_064643.10 | NP_497044.3 | 29 - 30 | 8435 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
