Assay Details
Gene Symbol: trip-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064699.5 | NP_497100.1 | 3 - 4 | 740 | 62 | |
GenBank mRNA | AF197575.1 | - | 3 - 4 | 750 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
