Assay Details
Gene Symbol: sel-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377920.3 | NP_001364559.1 | 16 - 17 | 4380 | 105 | |
NM_065538.6 | NP_497939.2 | 15 - 16 | 4245 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
