Assay Details
Gene Symbol: lips-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065540.3 | NP_497941.2 | 4 - 5 | 871 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
