Assay Details
Gene Symbol: Y32H12A.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372583.2 | NP_001360479.1 | 13 | 2193 | 87 | |
NM_001372585.2 | NP_001360485.1 | 12 - 13 | 2181 | 87 | ||
NM_001379709.1 | NP_001368729.1 | 12 - 13 | 2205 | 87 | ||
NM_001379710.2 | NP_001368730.1 | 13 - 14 | 2237 | 87 | ||
NM_065750.6 | NP_498151.2 | 12 - 13 | 2184 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
