Assay Details
Gene Symbol: osg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065771.7 | NP_498172.3 | 10 - 11 | 2136 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
