Assay Details
Gene Symbol: hmg-1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382889.1 | 1 - 2 | 255 | 68 | ||
NM_065974.8 | NP_498375.1 | 2 - 3 | 276 | 68 | ||
GenBank mRNA | AF056577.1 | - | 1 - 2 | 250 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
