Assay Details
Gene Symbol: hmg-1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027428.6 | NP_001022599.1 | 2 - 3 | 520 | 70 | |
NM_065974.8 | NP_498375.1 | 3 - 4 | 539 | 70 | ||
GenBank mRNA | AF056577.1 | - | 2 - 3 | 513 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
