Assay Details
Gene Symbol: hmg-1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027428.6 | NP_001022599.1 | 4 - 5 | 774 | 118 | |
NM_001382889.1 | 4 - 5 | 766 | 118 | |||
NM_065974.8 | NP_498375.1 | 5 - 6 | 793 | 118 | ||
GenBank mRNA | AF056577.1 | - | 4 - 5 | 767 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
