Assay Details
Gene Symbol: fbxl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379825.2 | NP_001367407.1 | 3 - 4 | 686 | 69 | |
NM_171210.7 | NP_741248.1 | 3 - 4 | 746 | 69 |
Gene Symbol: rfs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382934.1 | 3 - 4 | 745 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379825.2, NM_171210.7 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
