Assay Details
Gene Symbol: F59B2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379854.1 | NP_001366671.1 | 6 - 7 | 1807 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
