Assay Details
Gene Symbol: rpn-6.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268043.2 | NP_001254972.1 | 4 - 5 | 1130 | 92 | |
NM_001268044.3 | NP_001254973.1 | 2 - 3 | 495 | 92 |
Gene Symbol: F59B2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379854.1 | NP_001366671.1 | 4 - 5 | 1142 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379854.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
