Assay Details
Gene Symbol: T05G5.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268094.3 | NP_001255023.1 | 7 - 8 | 1421 | 80 | |
NM_001268095.3 | NP_001255024.1 | 2 - 3 | 298 | 80 | ||
NM_001268096.3 | NP_001255025.1 | 2 - 3 | 241 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
