Assay Details
Gene Symbol: mua-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027503.4 | NP_001022674.1 | 10 - 11 | 1636 | 73 | |
GenBank mRNA | AF139060.1 | - | 10 - 11 | 1636 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
