Assay Details
Gene Symbol: lonp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067176.4 | NP_499577.1 | 8 - 9 | 1547 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
