Assay Details
Gene Symbol: Y111B2A.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067232.7 | NP_499633.1 | 4 - 5 | 316 | 68 |
Gene Symbol: Y111B2A.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379955.2 | NP_001366979.1 | 3 - 4 | 618 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
