Assay Details
Gene Symbol: irld-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067258.2 | NP_499659.2 | 5 - 6 | 1326 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: irld-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067258.2 | NP_499659.2 | 5 - 6 | 1326 | 122 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |