Assay Details
Gene Symbol: Y37D8A.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067267.6 | NP_499668.1 | 8 - 9 | 1342 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
