Assay Details
Gene Symbol: Y37D8A.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067271.3 | NP_499672.1 | 4 - 5 | 442 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Y37D8A.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067271.3 | NP_499672.1 | 4 - 5 | 442 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() |