Assay Details
Gene Symbol: Y37D8A.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067271.3 | NP_499672.1 | 5 - 6 | 533 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Y37D8A.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067271.3 | NP_499672.1 | 5 - 6 | 533 | 57 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |