Assay Details
Gene Symbol: cla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307219.4 | NP_001294148.2 | 7 - 8 | 969 | 126 | |
NM_001307220.4 | NP_001294149.2 | 7 - 8 | 969 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
