Assay Details
Gene Symbol: ipla-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069096.3 | NP_501497.2 | 2 - 3 | 479 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
