Assay Details
Gene Symbol: C27B7.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268423.3 | NP_001255352.1 | 21 - 22 | 3148 | 119 | |
NR_056330.1 | 21 - 22 | 3138 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
