Assay Details
Gene Symbol: thk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268549.3 | NP_001255478.1 | 3 - 4 | 649 | 128 | |
NM_001383184.1 | 10 - 11 | 1695 | 128 | |||
GenBank mRNA | AY523517.1 | - | 4 - 5 | 691 | 128 |
Gene Symbol: rad-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028294.4 | NP_001023465.1 | 11 - 12 | 1839 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
