Assay Details
Gene Symbol: rad-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028294.4 | NP_001023465.1 | 6 - 7 | 784 | 76 | |
NM_001276842.3 | NP_001263771.1 | 4 - 5 | 685 | 76 | ||
NM_001383185.1 | 5 - 6 | 712 | 76 | |||
GenBank mRNA | AB011382.1 | - | 4 - 5 | 686 | 76 | |
AF061201.1 | - | 4 - 5 | 686 | 76 |
Gene Symbol: thk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383184.1 | 4 - 5 | 598 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383184.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
