Assay Details
Gene Symbol: sri-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276854.1 | NP_001263783.1 | 6 - 7 | 791 | 132 | |
NM_069642.1 | NP_502043.1 | 6 - 7 | 944 | 132 | ||
NR_131607.1 | 7 - 8 | 977 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
