Assay Details
Gene Symbol: cpt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047512.4 | NP_001040977.2 | 4 - 5 | 691 | 74 | |
NM_001047513.5 | NP_001040978.2 | 2 - 3 | 181 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
