Assay Details
Gene Symbol: nadk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067683.4 | NP_500084.2 | 1 - 2 | 110 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
