Assay Details
Gene Symbol: chat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028161.4 | NP_001023332.1 | 4 - 5 | 644 | 96 | |
NM_001047514.4 | NP_001040979.1 | 1 - 2 | 100 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
