Assay Details
Gene Symbol: cpi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068514.6 | NP_500915.1 | 1 - 2 | 166 | 67 | |
GenBank mRNA | AJ310669.1 | - | 1 - 2 | 103 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
