Assay Details
Gene Symbol: acl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069801.5 | NP_502202.2 | 2 - 3 | 274 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
