Assay Details
Gene Symbol: acl-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307027.3 | NP_001293956.1 | 1 - 2 | 125 | 141 | |
NM_069969.3 | NP_502370.1 | 4 - 5 | 972 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
