Assay Details
Gene Symbol: wdfy-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268727.2 | NP_001255656.1 | 12 - 13 | 1940 | 123 | |
NM_001268728.4 | NP_001255657.1 | 12 - 13 | 1921 | 123 | ||
NM_001268729.3 | NP_001255658.1 | 12 - 13 | 1942 | 123 | ||
NM_001268730.3 | NP_001255659.1 | 12 - 13 | 1854 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
