Assay Details
Gene Symbol: txt-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070045.3 | NP_502446.2 | 4 - 5 | 518 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: txt-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070045.3 | NP_502446.2 | 4 - 5 | 518 | 125 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |