Assay Details
Gene Symbol: Y51H4A.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380552.2 | NP_001366965.1 | 8 - 9 | 1476 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
