Assay Details
Gene Symbol: tank-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029135.6 | NP_001024306.1 | 6 - 7 | 5116 | 149 | |
NM_071000.8 | NP_503401.3 | 7 - 8 | 5230 | 149 | ||
GenBank mRNA | AF548287.1 | - | 6 - 7 | 5131 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
