Assay Details
Gene Symbol: tank-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029135.6 | NP_001024306.1 | 8 - 9 | 5894 | 75 | |
NM_071000.8 | NP_503401.3 | 9 - 10 | 6008 | 75 | ||
GenBank mRNA | AF548287.1 | - | 8 - 9 | 5909 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
