Assay Details
Gene Symbol: abhd-5.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071898.5 | NP_504299.2 | 4 - 5 | 753 | 57 |
Gene Symbol: abhd-5.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071896.6 | NP_504297.1 | 9 - 10 | 1963 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
