Assay Details
Gene Symbol: F08F3.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072032.7 | NP_504433.2 | 4 - 5 | 1082 | 90 |
Gene Symbol: F08F3.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182419.4 | NP_872219.2 | 4 - 5 | 987 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_182419.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
