Assay Details
Gene Symbol: acl-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028598.4 | NP_001023769.1 | 1 - 2 | 145 | 73 | |
NM_001028599.5 | NP_001023770.1 | 1 - 2 | 151 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001028599.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
