Assay Details
Gene Symbol: acl-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028598.4 | NP_001023769.1 | 7 - 8 | 1239 | 65 | |
NM_001028599.5 | NP_001023770.1 | 7 - 8 | 1241 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
