Assay Details
Gene Symbol: T05C3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072050.3 | NP_504451.3 | 7 - 8 | 1851 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: T05C3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072050.3 | NP_504451.3 | 7 - 8 | 1851 | 118 |
Set Membership: |
![]() ![]() ![]() ![]() |