Assay Details
Gene Symbol: fos-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038392.5 | NP_001033481.1 | 6 - 7 | 1286 | 69 | |
NM_001380651.1 | NP_001367492.1 | 7 - 8 | 1337 | 69 | ||
GenBank mRNA | AY835432.1 | - | 6 - 7 | 1293 | 69 | |
AY835433.1 | - | 6 - 7 | 1098 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
