Assay Details
Gene Symbol: ttn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029029.5 | NP_001024200.1 | 10 - 11 | 4292 | 64 | |
NM_001359765.3 | NP_001346723.1 | 2915 | 64 | |||
NM_001359766.3 | NP_001346716.1 | 2915 | 64 | |||
NM_001359767.3 | NP_001346722.1 | 4193 | 64 | |||
NM_001359768.3 | NP_001346715.1 | 4193 | 64 | |||
NM_001359777.3 | NP_001346724.1 | 9 - 10 | 3002 | 64 | ||
NM_001373034.1 | NP_001360072.1 | 4286 | 64 | |||
NM_001380656.1 | NP_001367914.1 | 2915 | 64 | |||
NM_001380657.1 | NP_001367912.1 | 3008 | 64 | |||
NM_001380658.2 | NP_001367913.1 | 4193 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
